Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD1 Recombinant Superclonal™ Antibody (8HCLC)

Catalog No. PI711181
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711181 Supplier Invitrogen™ Supplier No. 711181
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

RAD1 Recombinant Polyclonal Antibody for Western Blot, ICC/IF

Rad1 is a component of a heterotrimeric PCNA-like complex that contains Rad9 and Hus1 proteins. This complex is involved in the 9-1-1 cell cycle checkpoing response complex that plays a major role in DNA repair, by associating with Rad17 and several components of the PCNA-loading heteropentamer replication factor C. Mutants has been shown to partially restore radiation resistance and G2-checkpoint activity. It has also been shown to possess exonuclease activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 8HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAD1
Gene Accession No. O60671
Gene Alias cell cycle checkpoint protein Hrad1; cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; mRAD1; RAD1; RAD1 checkpoint clamp component; RAD1 checkpoint DNA exonuclease; RAD1 homolog; RAD1 homolog (S. pombe); rad1-like DNA damage checkpoint protein; REC1
Gene Symbols RAD1
Host Species Rabbit
Immunogen Protein corresponding to Human RAD1 (aa 133-282).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.