Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD50 Monoclonal Antibody (13B3)
GREENER_CHOICE

Catalog No. 01950107 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-107 Supplier Invitrogen™ Supplier No. MA123269
  • $696.00
    $550.65 / Each of 1

    Save $145.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Recommended positive controls: HeLa, HeLa nuclear extract.

Rad50-13B3 recognizes human Rad50, a 153a protein involved in DNA double strand break repair. Rad50 is associated with Mre11 and p95 (Nibrin) to form a multiprotein complex involved in the double strand break repair process. Upon DNA damage, Rad50 and Mre11 redistribute and are co-localized within the nucleus of damaged cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD50
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Proximity Ligation Assay, Immunocytochemistry
Classification Monoclonal
Clone 13B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RAD50
Gene Accession No. P70388, Q92878, Q9JIL8
Gene Alias 153 kDa protein; DNA repair protein RAD50; hRad50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; Rad50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W
Gene Symbols RAD50
Host Species Mouse
Immunogen Amino acids 1-425 of Rad50 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10111, 19360, 64012
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.