Learn More
Diagnostic Biosystems RAD51 Rad51
Supplier:  Diagnostic Biosystems MOB32405
Encompass_Preferred
This antibody is specific to a 37-43 kDa protein known as Rad51. Rad51 is similar to RecA. Rad51 promotes homologous pairing and strand exchange within a regular nucleoprotein filament. Rad51 has been suggested to be involved in homologous recombination and in cell proliferation regulation.
Catalog No. 50-238-5382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.