Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534905 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534905 Supplier Invitrogen™ Supplier No. PA534905
  • $624.00
    $494.65 / Each of 1

    Save $129.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: yeast. Predicted reactivity: Yeast (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Rad51
Gene Accession No. P25454
Gene Alias AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754
Gene Symbols Rad51
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Saccharomyces cerevisiae S288c Rad51. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856831
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.