Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD51C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565401 Supplier Invitrogen™ Supplier No. PA565401
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRGKTFRFEMQ RDLVSFPLSP AVRVKLVSAG FQTAEELLEV KPSELSKEVG ISKAEALETL QIIRRECLTN KPRYAGTSES HKKCTALELL EQEHTQGFII TFCSALDDIL GGGVPLMKTT EICGAPGVG Highest antigen sequence identity to the following orthologs - mouse 79%, rat 82%.

ZFPM1 is a transcription regulator that plays an essential role in erythroid and megakaryocytic cell differentiation. ZFPM1 is an essential cofactor that acts via the formation of a heterodimer with transcription factors of the GATA family GATA1, GATA2 and GATA3. Such heterodimer can both activate or repress transcriptional activity, depending on the cell and promoter context. The heterodimer formed with GATA proteins is essential to activate expression of genes such as NFE2, ITGA2B, alpha- and beta-globin, while it represses expression of KLF1. ZFPM1 may be involved in regulation of some genes in gonads. ZFPM1 may also be involved in cardiac development, in a non-redundant way with ZFPM2/FOG2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAD51C
Gene Accession No. O43502
Gene Alias BROVCA3; DNA repair protein RAD51 homolog 3; FANCO; MGC104277; R51H3; RAD51 homolog C; RAD51 paralog C; RAD51C; RAD51L2; RAD51-like protein 2; RGD1563765; yeast RAD51 homolog 3
Gene Symbols RAD51C
Host Species Rabbit
Immunogen Recombinant Human RAD51C. Recombinant protein control fragment (Product #RP-106082).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5889
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.