Learn More
Alkali Scientific Radiant™ 2X GC-Long Mastermix 400 Reactions 10 x 1mL
SDP
Supplier:  Alkali Scientific C559
Encompass_Preferred
Radiant™ GC-Long Hot Start 2x Master Mix has been engineered for the amplification of extremely difficult templates and long amplicons up to35kb. Radiant™ GC-Long Hot Start 2x Master Mix possesses 5´-3´ DNA polymerase and 3´-5´ proofreading exonuclease activities with an errorrate of 4.55 x 10-7. The polymerase is ideal for cloning applications, library construction, screening and genotyping of problematic GC-rich andAT-rich sequences. Radiant™ GC-Long Hot Start 2x Master Mix Polymerase is engineered for robust, superior high-fidelity PCR in comparisonwith standard proof-reading enzymes such as pfu DNA Polymerase. Several point-mutations combined with a new-generation bufferformulation provide for significantly improved processivity for faster cycling, greater sensitivity and less inhibition with crude DNA samples.
Catalog No. 50-238-9565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.