Learn More
United Scientific Supplies Radiation Can Kit
SDPSCI_ED
Catalog No. S07397 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Lamp Assembly
Radiation Can Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07397 Radiation Can Kit
S07396 Lamp Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S07397 Supplier United Scientific Supplies Supplier No. RDCNKIT
Add to Cart
Add to Cart
Includes
Lamp assembly with reflector and stand (bulb not included), thermometer, one smooth metal can, one black metal can, thermometer.
Demonstrate how two identically sized cans absorb and radiate energy differently.
  • Includes two cans: one smooth metal can w/screw cap and other black metal can
  • Thermal radiation emitted by two cans cools or heats water at different rates
Order Info
Lamp assembly with stand is also sold separately.
Specifications
For Use With (Application) For demonstrate how two identically sized cans absorb and radiate energy differently
Includes Lamp assembly with reflector and stand (bulb not included), thermometer, one smooth Metal can, one black Metal can, thermometer
Product Type Radiation Can Kit