Learn More
Thermo Scientific™ RadHalo FM Fixed Spectroscopic Area Monitors
Description
- Rapid dose rate response
- Immediate accurate identification
- Autonomous operation anywhere
- Durable design
- Secure transmissions
- Detector Fusion Capable
Specifications
Specifications
| Detector Type | Spectroscopic Detector: 3 x 3 in. NaI |
| Description | RadHalo FM-SE |
| Communications | Wi-Fi, Remote Satellite Transceiver for Reachback |
| Includes | Fixed with extended satellite |