Learn More
Research Products International Corp Radicicol [Humicola Fuscoatra], 5 Milligrams
SDP
Supplier:  Research Products International Corp R200200.005
Encompass_Preferred
Radicicol is a naturally occurring antibiotic and antifungal compound . It is known for its ability to inhibit the activity of heat shock protein 90 (Hsp90), a molecular chaperone that plays a crucial role in the folding and stabilization of various client proteins involved in cell signaling and regulation. 
Radicicol is often used as a tool to study Hsp90-dependent processes and the functions of client proteins. Researchers use radicicol to investigate the impact of inhibiting Hsp90 on cellular pathways, protein folding, and cell viability. This compound is particularly valuable in cancer research, as many oncogenic proteins are dependent on Hsp90 for proper folding and stability.
Catalog No. 50-488-977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.