Learn More
MilliporeSigma™ Supelco™ radiello™ Anesthetic Gases and Vapor Sampler
Catalog No. 111008561 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Sampler; 1 permeative body. 1 support plate. 1 vertical adapter. 1 adsorbing cartridge
Catalog No.
11-100-8561 Sampler; 1 permeative body. 1 support plate. 1 vertical adapter. 1 adsorbing cartridge
1 options
Catalog No. 11-100-8561 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD125
Add to Cart
Add to Cart
Designed for sampling of nitrous oxide, isofluorane, ethrane, halothane, and sevorane in surgical theaters
Parts for one complete sampler are packed separately in one sealed and sterile bag. Pack of 10 sterile bags. Each sterile bag contains:. 1 permeative body. 1 support plate. 1 vertical adapter. 1 adsorbing cartridge
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Sampling of Nitrous Oxide, Isofluorane, Ethrane, Halothane, and Sevorane in Surgical Theaters
Includes Parts for one complete sampler are packed separately in one sealed and sterile bag. Pk. of 10 sterile bags.
Type Air Sampler
radiello is a trademark of Institi Clinici Scientifici Maugeri