Learn More
MilliporeSigma™ Supelco™ radiello™ Empty Storage Tubes, Glass (with stopper)
Catalog No. 111009779 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Type:
2.8 mL Tube
Catalog No. Type
11-100-9779 2.8 mL Tube
1 options
Catalog No. 11-100-9779 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD1991
Add to Cart
Add to Cart
Used for storing radiello™ Cartridge Adsorbents before and after diffusive sampling
TRUSTED_SUSTAINABILITY
Specifications
Material Glass (with Stopper)
For Use With (Application) Air Monitoring
Quantity 100/Pk.
Type 2.8 mL Tube
radiello is a trademark of Institi Clinici Scientifici Maugeri