Learn More
MilliporeSigma™ Supelco™ radiello™ Empty Storage Tubes, Polypropylene (with stopper)
Catalog No. 111010214 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
11-101-0214 100/Pk.
1 options
Catalog No. 11-101-0214 Supplier MilliporeSigma™ Supelco™ Supplier No. RAD1992
Add to Cart
Add to Cart
Used for storing radiello™ Cartridge Adsorbents before and after diffusive sampling
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene (with Stopper)
Quantity 100/Pk.
Type 12 mL Tube
radiello is a trademark of Institi Clinici Scientifici Maugeri