Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys Reactor-Ready™ Flex Pro Lab Reactor
SDP

Catalog No. 13990611 Shop All Heidolph North America Products
Change view
Click to view available options
Volume (Metric):
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-990-611 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-990-611 Supplier Heidolph™ Supplier No. 015950102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-volume lab reaction station for glass reaction vessels from 250 mL to 5 L. Heidolph Radley Reactor-Ready Flex is designed as a universal reactor work station that can be used for different vessel sizes and different experiments.

  • Radleys Reactor-Ready Flex is designed as a universal reactor workstation with a range of easily interchangeable vessels from 100 mL to 5 liters. This can be configured to suit the chemistry and scale needed for each project.
  • Ideal for precise temperature control (-70 °C to +230 °C), stirring, and dosing, Reactor-Ready Flex allows chemists to begin process development earlier, with added confidence and safety.
  • Includes complete turnkey frame
  • Framework: Starter, Includes Drain-down Manifold Kit: No, Stirrer Alignment: Manual, Vessel Clamp: Separate
  • Starter:
  • Core functionality for limited budgets
  • No drain-down manifolds
  • Compact stirrer support for manual alignment
  • Starter can be upgraded to Pro with the addition of the drain-down manifold kit
  • Pro:
  • With drain-down manifolds for rapid, mess-free vessel exchange
  • Manifolds take the weight of the hoses
  • I-Beam Stirrer Support - easy alignment on 3 axes
  • Configure Reactor-Ready Flex with the help of your Fisher Scientific sales rep
  • 1) Select Framework - Starter or Pro
  • 2) Select Vessel Kits
  • 3) Select Overhead Stirrer
  • 4) Select Circulator and Accessories
  • 5) Select Software Control
  • 6) Select Software Modules
  • 7) Select Glassware and Accessories

Specifications

Volume (Metric) 5 L
For Use With (Equipment) Framework only - paired with vessel kits, overhead stirrer, circulator, glassware and accessories and optional software
Includes Turnkey framework only - single jacketed or vacuum jacketed vessels available separately
Product Type Lab Reactor
Warranty 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.