Learn More
Heidolph™ Radleys Reactor-Ready™ Mini Lab Reactor
SDP
Catalog No. 13889881 Shop All Heidolph North America Products
Change view
Click to view available options
Model:
Pro
Starter
Includes:
Draindown Manifold Kit
No Draindown Manifold Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
13-889-881 Pro Draindown Manifold Kit
13-889-880 Starter No Draindown Manifold Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-889-881 Supplier Heidolph™ Supplier No. 015920012
Only null left
Add to Cart
Add to Cart
A compact benchtop jacketed lab reactor designed for small‐scale chemistry, offering the same precision, control and flexibility as a full-size Reactor-Ready system
  • Radleys Reactor-Ready Mini is a small benchtop jacketed lab reactor that accepts vessels from 50 to 500 ml.
  • Designed for small-scale chemistry, it offers the precision, control, and flexibility of a full-size reactor-ready system in a compact, space-saving footprint.
  • Ideal for precise temperature control (-70°C to +230°C), stirring, and dosing, Reactor-Ready Mini allows chemists to begin process development earlier, with added confidence and safety.
Includes complete turnkey frame
Starter:
  • For more limited budgets, connect your vessel directly to your circulator
  • No draindown manifolds
  • The starter can be upgraded to Pro with the addition of the draindown manifold kit
Pro:
  • With draindown manifolds for rapid, mess-free vessel exchange
  • Manifolds take the weight of the hoses
Configure Reactor-Ready Mini with the help of your Fisher Scientific sales rep
  • Select Framework - Starter or Pro
  • Select Vessel Kits
  • Select Overhead Stirrer
  • Select Circulator and Accessories
  • Select Software Control
  • Select Software Modules
  • Select Glassware and Accessories
Specifications
Volume (Metric) 500 mL
For Use With (Equipment) Overhead Stirrer, Circulator, Glassware and Accessories
Includes Draindown Manifold Kit
Product Type Lab Reactor
Warranty 1 Year
Model Pro