Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Raf-1 RBD Agarose Beads, Liquid, 400 μg

Catalog No. 502588463
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-8463 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-8463 Supplier Abcam Supplier No. AB211176400UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Affinity purification reagent supplied as a liquid in a 50% glycerol storage buffer.

  • Form: Liquid
  • Storage buffer: Constituents: 50% Glycerol (glycerin, glycerine)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Protein-based Substrate
Target Molecule Ras
Format Beads
Form Liquid
Buffer 50% glycerol
Formulation 50% Glycerol
Quantity 400 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.