Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein

Catalog No. PV3805
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3805 10 μg
PR7307A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3805 Supplier Invitrogen™ Supplier No. PV3805
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic, Mutant.

RAF1 (cRAF) is a MAP kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated RAF1 can phosphorylate and activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylates and activates the serine/threonine specific protein kinases ERK1 and ERK2. The RAF1 Y340D Y341D mutant is constitutively active. RAF1 functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 2mM DTT, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
Molecular Weight (g/mol) 66.4 kDa
Name Human RAF1 (cRAF) [Y340D, Y341D], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name c-Raf (Y340D, Y341D Mutant)
Kinase Family RAF Kinase Family
Protein Length 306-648; Y340D, Y341D
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 306-648; Y340D, Y341D
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry RAF1 (cRAF) Y340D Y341D
Target Gene v-raf-1 murine leukemia viral oncogene homolog 1
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic, Mutant
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Purity or Quality Grade 80% by SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.