Learn More
Abcam Raffinose Pentahydrate, Trisaccharide, >98% Purity, 25 g
Description
- Molecular Weight: 594.5 g/mol
- Purity: >98%
- Composition: Trisaccharide composed of galactose, fructose, and glucose
- Hydrolysis: Hydrolysed to D-galactose and sucrose by D-galactosidase
- Storage buffer: 100% Raffinose pentahydrate
- CAS Number: 17629-30-0
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Powder |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% Raffinose pentahydrate |
| Formulation | 100% Raffinose pentahydrate |
| Quantity | 25 g |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.