Learn More
Medchemexpress LLC HY-17598 500mg Medchemexpress, Rafoxanide CAS:22662-39-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17598500MG
Encompass_Preferred
Medchemexpress, HY-17598 500mg Rafoxanide CAS:22662-39-1 Rafoxanide is an orally active salicylanilide anthelmintic agent. Rafoxanide is an antiparasitic agent and can be used for the control of infestation with Hemonchus species and Fasciola species in sheep and cattle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1998
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.