Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Rainbow™ Molecular Weight Markers

Catalog No. p-3667614 Shop All Cytiva Products
Change view
Click to view available options
Concentration:
1 mg/mL
1.5 mg/mL
2 mg/mL
Range:
12000 to 225000Mr
3500 to 38000Mr
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Range
45-001-593 1 mg/mL 3500 to 38000Mr
45-001-592 1.5 mg/mL 12000 to 225000Mr
45-001-591 2 mg/mL 12000 to 225000Mr
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-593 Supplier Cytiva Supplier No. RPN755E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining. Cytiva Life Sciences™ Rainbow™ Molecular Weight Markers use bright, distinctive colors to allow easier confirmation of transfer to blotting membranes and orientation.

Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination.

  • High intensity, prestained multicolored bands allow distinction of closely related proteins
  • Full-Range Rainbow Markers (Mr 12,000 to 225,000) use ten separate proteins with six different colors
  • High-Range Rainbow Markers (Mr 12,000 to 225,000) use eight separate proteins with six different colors
  • Low-Range Rainbow Markers (Mr 3500 to 38,000) use seven separate proteins with five different colors
  • For use on SDS-PAGE

  • The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
  • Markers are a mixture of individually colored proteins that are combined to produce bands of equal color intensity. Approximate protein concentration is 1 to 2 mg/mL, depending on the product
  • Each 250μL pack size is sufficient for use with 50 minigels (10 × 8cm) or 25 large gels (20 × 20cm)

TRUSTED_SUSTAINABILITY

Specifications

Description Rainbow molecular weight markers enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels.
Color Multi
Concentration 1 mg/mL
Form Liquid
Formulation Supplied ready to use in 305% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent
For Use With (Application) For industrial applications: Analytical reagent, research Use seven separate proteins with five different colors
Molecular Weight (g/mol) Mr 3 5 00
Quantity 250 μL
Range 3500 to 38000Mr
Storage Requirements -15 to -30°C
Sufficient For 50 Minigels or 25 large gels
Product Type Protein Molecular Weight Marker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.