Learn More
Mascon Medical Mascon Medical™ Rainin Pipette Tips, 10ul, Sterile, Filtered, 96EA/Rack, 10 Racks/Box, 5 Boxes/Case
SDP
Supplier:  Mascon Medical FSRAI10S
Encompass_Preferred
Mascon Medical’s 10ul Rainin Compatible Pipette Tips offer superior performance and contamination-free sample transfer. Designed to seamlessly fit Rainin pipettes, they ensure a secure seal and easy ejection. With low retention, these transparent polypropylene tips minimize residue for reliable results.Measuring 49.3mm, they provide precise handling and withstand -40℃ to 100℃. Each pack contains 96 tips, with 10 racks per box and 5 boxes per carton, ensuring an ample supply.Sterile and filtered, these tips prevent cross-contamination during molecular biology and genetics experiments. The advanced mold design and smooth wall minimize residue. Their burr-free small mouth and high concentricity prevent cross-contamination.With a filter to protect against corrosive liquids and meeting ISO13485 standards, Mascon Medical™ ensures quality. Compatible with Rainin pipettes, these tips deliver optimal performance. Trust Mascon Medical™ for reliable pipetting.
Catalog No. NC2458322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.