Learn More
National Safety Apparel™ Rainwear Waist Pants Polyurethane Coated FR Weft
SDP
Catalog No. 170012234 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2234 4X-Large
17-001-2229 Small
17-001-2230 Medium
17-001-2231 Large
17-001-2232 2X-Large
17-001-2233 3X-Large
17-001-2235 X-Large
17-001-2236 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2234 Supplier National Safety Apparel™ Supplier No. R50RL144X
Add to Cart
Add to Cart
Specifications
Size 4X-Large
Gender Unisex
Material Polyurethane-coated FR Cotton
Color Fluorescent Yellow
Ankle Style Open
Seam Type Welded and Taped
Closure Type Buckle
Disposable Reusable
Arc Flash Rating 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange)
Certifications/Compliance CAT 2, NFPA 70 E, ANSI/ISEA 107 Class E, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Sterility Non-sterile