Learn More
Tecniplast™ Raised Wire Floors
Catalog No. 012886C Shop All Tecniplast USA Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Fits cage size C
Fits cage size F
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-288-6C Fits cage size C
01-288-6F Fits cage size F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-288-6C Supplier Tecniplast™ Supplier No. 1264C150
Add to Cart
Add to Cart
Raised bottom grid can be dropped into solid bottom cage to separate animals from their waste.
  • 30mm elevation separates animals from bedding and waste
  • Convenient handle design for easy insertion and removal
  • Made of AISI 304 Stainless steel mesh
Specifications
Product Type Wire Floor
Material Stainless Steel (304)
For Use With (Application) Seprate animals from their waste
For Use With (Equipment) Fits cage size C
Width (English) 8.26 in.
Height (English) 5.5 in.
Height (Metric) 14 cm