Learn More
Abcam Raji Membrane Extract Lysate, Liquid, WB, 100 μg
Description
- Applications: WB
- Form: Liquid
- Storage buffer: Constituents: HEPES, Magnesium chloride, Potassium chloride, Sucrose, 10% Glycerol, 2% Sodium lauryl sulfate, 1.2% 2-Mercaptoethanol, 1% 4-Nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Detection Location | In-Blot Detection |
| Format | Tube |
| Form | Liquid |
| Buffer | HEPES, magnesium chloride, potassium chloride, sucrose, 10% glycerol, 2% sodium lauryl sulfate, 1.2% 2-mercaptoethanol, 1% 4-nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione |
| Reactivity | Human |
| Target Molecule | Raji Cells (Human) |
| Cross Reactivity | Human |
| Formulation | HEPES, magnesium chloride, potassium chloride, sucrose, 10% glycerol, 2% sodium lauryl sulfate, 1.2% 2-mercaptoethanol, 1% 4-nonylphenol, branched, ethoxylated, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione |
| Quantity | 100 μg |
| For Use With (Application) | Western Blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.