Learn More
Stoelting™ Rake Retractor
SDP
Stoelting™ Rake Retractor can be used to actively separate the edges of a surgical incision or wound or hold back underlying organs and tissues so that body parts under the incision may be accessed.
$154.80 - $169.60
Specifications
For Use With (Application) Neuroscience
Length (English) 6 in.
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-001-426
SDP
View Documents
Stoelting™
5212625
2 blunt prongs
Each for $154.80
Only null left
Add to Cart
10-001-427
SDP
View Documents
Stoelting™
5212626
3 blunt prongs
Each for $169.60
Only null left
Add to Cart
Description
Description
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts.
  • Blunt retractors are recommended when iatrogenic injury is a concern and the main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
Specifications
Neuroscience
6 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.