Description
Includes
- Ral Activation Assay Buffer, 5X
- 100X GTPγS, 10mM
- 100X GDP, 100mM
- Anti-Ral-A
- Ral Assay Reagent (Ral BP1, agarose)
Specifications
Specifications
| For Use With (Application) | Affinity Binding Assay |
| Quantity | 30 Assays/Kit |
| Type | Activation Assay Kit |