Learn More
Medchemexpress LLC HY-101437 50mg Medchemexpress, Ralfinamide CAS:133865-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10143750MG
Encompass_Preferred
Medchemexpress, HY-101437 50mg Ralfinamide CAS:133865-88-0 Ralfinamide (FCE-26742A) is an orally available Na + blocker derived from α-aminoamide, with function of suppressing pain [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.