Learn More
Abcam RalGDS RBD Agarose Beads, Liquid, 400 μg
Description
- Form: Liquid
- Storage buffer: 50% Glycerol (glycerin, glycerine)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 50% glycerol |
| Format | Beads |
| Form | Liquid |
| Target Molecule | Ral (GTP-bound) |
| Formulation | 50% Glycerol |
| Quantity | 400 μg |
| For Use With (Application) | Functional studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.