Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALGPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569173 Supplier Invitrogen™ Supplier No. PA569173
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 76%.

RALGPS1 gene ontology annotations related to this gene include regulation of Ral protein signal transduction; small GTPase mediated signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALGPS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RALGPS1
Gene Accession No. Q5JS13
Gene Alias 5830418G11Rik; AI853783; AI854138; Kiaa0351; mKIAA0351; Ral GEF with PH domain and SH3 binding motif 1; ral GEF with PH domain and SH3-binding motif 1; Ral guanine nucleotide exchange factor 2; Ral guanine nucleotide exchange factor RalGPS1A; ralA exchange factor RalGPS1; RalGEF 2; RALGEF2; RALGPS1; Ralgps1a; ras-specific guanine nucleotide-releasing factor RalGPS1; si:dkey-191c17.1; zgc:165535
Gene Symbols RALGPS1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RALGPS1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9649
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.