Learn More
RAM Scientific Safet-T-Fil™ Blood Gas Capillaries
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes through the same capillary action as glass without the risk of breakage. Capillary tubes are available untreated or heparinized.
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Replaces glass Natelson, Caraway, or mylar-coated glass blood gas capillary tubes
- Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
- Capillary tubes are available untreated or heparinized
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- Optional end caps minimize the risk of blood spillage and allow for easier handling
- Optional mixing fleas and magnetic rings (mixing components) allow for uniform mixing to prevent clotting
- Packaging labels are marked with Lot Number and Expiration Date
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
| Color | Clear |
| Product Type | Blood Gas Tube |
| Additive | Balanced Heparin |
| Material | Plastic |
| Sterility | Non-sterile |
| Capacity (Metric) | 230 μL |
| For Use With (Application) | Intended for blood gas analysis, tests for multiple analytes, or as a transfer device for PKU and POC testing and coated with Balanced Heparin anticoagulant to prevent clotting. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.