Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Euramco Ramfan™ Intrinsically Safe Electric Blower
SDP

Catalog No. 19817726
Click to view available options
:
Blower/Exhaust Fan, Intrinsically saf

Catalog No. 19-817-726

Supplier: Euramco EB7201XX

Includes

30 ft. (9.1m) power cord (plug sold separately).

Blow or exhaust hazardous/explosive gases safely

  • High-pressure axial blower for hazardous locations
  • Intrinsically safe motor, 100% duty cycle TEAO (Totally Enclosed Air Over)
  • Tough, chemical-resistant ABS housing, carbon-filled to conduct static charges to ground
  • IP65 rain tested switch enclosure
  • Integral duct adapters
  • Moves 2500cfm of free air with 12 in. fan driven by 3/4hp motor
  • Uses 12 in. (30.5cm) duct
  • 115/230VAC 50/60Hz, single phase
  • Plug not included

Certifications

Motor UL Class I Group D and UL Class II Group E, F, G certified.

Compliance

Meet or exceed all applicable OSHA safety standards. Noise level: 89dB.

Warranty and Services

One year

Specifications

Color Black
Depth (English) 16 in.
Depth (Metric) 40.64 cm
Certifications/Compliance ATEX, UL
Diameter (English) Fan 16 in.
Amperage 7.2 A
Diameter (Metric) Fan 40.64 cm
Product Type Intrinsically Safe Blower/Exhauster
Hertz 50/60 Hz
Voltage 115/230 V
Electrical Requirements 115/230 V, 50/60 Hz
Flow Rate 2500 cfm
Height (English) 16 in.
Horsepower 0.75 hp
Includes 30 ft. Power cord
Length (English) Duct 12 in.
Length (Metric) Duct 30.48 cm
Height (Metric) 40.64 cm
Material Conductive ABS
Noise 89 dB
Width (Metric) 38.1 cm
Weight (English) 44 lb.
Weight (Metric) 19.95 kg
Width (English) 15 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.