Learn More
Apexbio Technology LLC Ramiprilat 87269-97-4 1mg
SDP
Supplier:  Apexbio Technology LLC C34921
Encompass_Preferred
Ramiprilat (CAS 87269-97-4) is an active metabolite of the ACE inhibitor prodrug ramipril It selectively inhibits angiotensin-converting enzyme (ACE) thereby reducing the formation of angiotensin II a peptide involved in vasoconstriction and aldosterone secretion In human heart ramiprilat demonstrates high binding affinity for ACE (pKi 9 08) In vitro studies indicate that ramiprilat modulates B2 kinin receptor dynamics and decreases [3H]bradykinin binding in endothelial cell membranes In animal models ACE inhibition with ramiprilat ameliorates diabetic renal injury and hypertension Clinically ramiprilat is associated with decreased risk of cardiovascular events supporting its value in cardiovascular and renal research
Catalog No. 50-000-91387
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.