Learn More
AdipoGen Ramoplanin A2
SDP
Supplier:  AdipoGen AGCN20318M001
Encompass_Preferred
Chemical. CAS 81988-88-7 A2. Formula C119H154ClN21O40. MW 2554.1. Isolated from Actinoplanes sp. Glycolipodepsipeptide antibiotic. Complex of structurally related molecules A1, A2 and A3, with ramoplanin A2 as the primary component. Antibacterial and antiviral agent with activity against aerobic and anaerobic Gram-positive bacteria such as Clostridium difficile and antibiotic-resistant enterococci. Inhibits cell wall synthesis and consequently bacterial growth by forming a complex with lipid intermediate II Lipid II, a key intermediate in peptidoglycan biosynthesis.
Catalog No. 50-256-2604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.