Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142664
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142664 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142664 Supplier Invitrogen™ Supplier No. PA5142664
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RAN
Gene Accession No. P62826, P62827, P62828
Gene Alias AAF30287; androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Chromosome stability protein 17; CNR1; CST17; Dmel\CG1404; Dmel_CG1404; dran; Genetic suppressor of PRP20-1; GM14354p; gsp1; GTPase Ran; GTPase Ran homolog; GTP-binding nuclear protein GSP1/CNR1; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; L8003.19; LD32416p; LD40852p; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; Ran GTPase GSP1; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; ras-related nuclear protein; tc4; XELAEV_18007741mg; xran; YLR293C
Gene Symbols RAN
Host Species Goat
Immunogen Peptide with sequence C-YEHDLEVAQTTALP (aa 197-210).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19384, 5901, 84509
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.