Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1080 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1080 Supplier Invitrogen™ Supplier No. PA1080
  • $619.55
    $481.65 / Each of 1

    Save $137.90

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-080 detects Ran binding protein 1 (RanBP1) from human and mouse cells. PA1-080 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~30 kDa protein representing RanBP1 from HeLa cell extract. The PA1-080 immunogen is a synthetic peptide corresponding to residues M(1) A A A K D T H E D H D T S T E N T(18) C of human RanBP1. The PA1-080 immunizing peptide (Cat. # PEP-128) is available for use in neutralization and control experiments.

Ran/TC4-binding protein, RanBP1, interacts specifically with GTP-charged RAN. RANBP1 encodes a 23-kD protein that binds to RAN complexed with GTP but not GDP. RANBP1 does not activate GTPase activity of RAN but does markedly increase GTP hydrolysis by the RanGTPase-activating protein (RanGAP1). The RANBP1 cDNA encodes a 201-amino acid protein that is 92% similar to its mouse homolog. In both mammalian cells and in yeast, RANBP1 acts as a negative regulator of RCC1 by inhibiting RCC1-stimulated guanine nucleotide release from RAN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene RANBP1
Gene Accession No. P34022, P43487
Gene Alias HpaII tiny fragments locus 9A; hpaII tiny fragments locus 9a protein; Htf9a; Htf9-a; Htf9-a/RanBP1; RAN binding protein 1; ran-binding protein 1; RANBP1; RANG; Ran-specific GTPase-activating protein; testis secretory sperm-binding protein Li 221n
Gene Symbols RANBP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) A A A K D T H E D H D T S T E N T(18) C of human RanBP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19385, 5902
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.