Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP2 Monoclonal Antibody (2E1)
GREENER_CHOICE

Catalog No. PIMA1847
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1847 Supplier Invitrogen™ Supplier No. MA1847
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-847 detects Nup358/RanBP2 from human samples.

RanBP2 interacts with the small GTP-binding protein Ran and forms a large multiprotein complex involved in many biological processes such as mRNA processing, nuclear transport, cell cycle control and postmitotic nuclear reassembly. The protein is identified by western blotting as an apparent molecular mass of 360 kD. It is involved in nucleation of microtubule and control of cell growth by interacting with many protein factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene RANBP2
Gene Accession No. P49792
Gene Alias 358 kDa nucleoporin; A430087B05Rik; acute necrotizing encephalopathy 1 (autosomal dominant); ADANE; AI256741; ANE1; E3 SUMO-protein ligase RanBP2; E3 SUMO-protein transferase RanBP2; IIAE3; Nuclear pore complex protein Nup358; nucleoporin 358; nucleoporin Nup358; NUP358; P270; RAN binding protein 2; ran-binding protein 2; RANBP2; RBP2; retina-specific cyclophilin; RGD1560047; spliced variant with Ran-binding and cyclophilin domains; transformation-related protein 2; TRP1; TRP2
Gene Symbols RANBP2
Host Species Mouse
Immunogen Recombinant protein corresponding to 150 N-terminal residues of human Nup358.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5903
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.