Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14995 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14995 Supplier Invitrogen™ Supplier No. PA14995
  • $746.00
    $568.65 / Each of 1

    Save $177.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-4995 detects RanBP2 from human samples. PA1-4995 has been successfully used in Western blot, immunoprecimmunoprecipitationitation, and immunohistochemistry applications. The PA1-4995 immunogen is synthetic peptide corresponding to residues SKAPKSGFEGMFTKKE, repeated 3 times.

RanBP2 interacts with the small GTP-binding protein Ran and forms a large multiprotein complex involved in many biological processes such as mRNA processing, nuclear transport, cell cycle control and postmitotic nuclear reassembly. The protein is identified by western blotting as an apparent molecular mass of 360 kD. It is involved in nucleation of microtubule and control of cell growth by interacting with many protein factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Whole serum with 0.09% sodium azide
Gene RANBP2
Gene Accession No. P49792, Q9ERU9
Gene Alias 358 kDa nucleoporin; A430087B05Rik; acute necrotizing encephalopathy 1 (autosomal dominant); ADANE; AI256741; ANE1; E3 SUMO-protein ligase RanBP2; E3 SUMO-protein transferase RanBP2; IIAE3; Nuclear pore complex protein Nup358; nucleoporin 358; nucleoporin Nup358; NUP358; P270; RAN binding protein 2; ran-binding protein 2; RANBP2; RBP2; retina-specific cyclophilin; RGD1560047; spliced variant with Ran-binding and cyclophilin domains; transformation-related protein 2; TRP1; TRP2
Gene Symbols RANBP2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues SKAPKSGFEGMFTKKE, repeated 3 times.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19386, 294429, 5903
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.