Learn More
Description
PA1-082 detects Ran binding protein 2 (RanBP2) from human and bovine retina. PA1-082 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~360 kDa protein representing RanBP2 from bovine retinal extracts. Immunohistochemical staining of bovine retina with PA1-082 results in cytoplasmic staining of cells within the inner nuclear layer as well as the ganglion cells. The PA1-082 immunogen is a synthetic peptide corresponding to residues M(1) R R S K A D V E R Y I A S V Q G S(18) of human RanBP2. This sequence is completely conserved between human and mouse. The PA1-082 immunizing peptide (Cat. # PEP-137) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | RANBP2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | RANBP2 |
| Gene Accession No. | P48820, P49792 |
| Gene Alias | 358 kDa nucleoporin; A430087B05Rik; acute necrotizing encephalopathy 1 (autosomal dominant); ADANE; AI256741; ANE1; E3 SUMO-protein ligase RanBP2; E3 SUMO-protein transferase RanBP2; IIAE3; Nuclear pore complex protein Nup358; nucleoporin 358; nucleoporin Nup358; NUP358; P270; RAN binding protein 2; ran-binding protein 2; RANBP2; RBP2; retina-specific cyclophilin; RGD1560047; spliced variant with Ran-binding and cyclophilin domains; transformation-related protein 2; TRP1; TRP2 |
| Gene Symbols | RANBP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
