Learn More
Enzo Life Sciences RanBP2 Delta FG (human), (recombinant) (GST-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW94550100
Encompass_Preferred
RanBP2 directly interacts with the E2 enzyme Ubc9 and strongly enhances SUMO-1 transfer from Ubc9 to the SUMO-1 target Sp100. The SUMO1 E3 ligase activity is contained within a 33kDa domain of RanBP2 that lacks RING finger motifs. RanBP2 directly interacts with the E2 enzyme Ubc9 and strongly enhances SUMO-1 transfer from Ubc9 to the SUMO-1 target Sp100. The SUMO1 E3 ligase activity is contained within a 33kDa domain of RanBP2 that lacks RING finger motifs. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 20mM HEPES, pH7.3 containing 110mM potassium acetate, 2mM magnesium acetate, 0.8mM EGTA, 1mM DTT. Source: Produced in E. coli. RanBP2ΔFG (aa 2553-2838) is fused to a GST-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2315
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.