Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EvenCoat Goat Anti-Mouse IgG Coated Plates

Catalog No. CP001
Change view
Click to view available options
Quantity:
15/Pk.
5/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CP001 5/Pk.
CP002 15/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. CP001 Supplier R&D Systems™ Supplier No. CP001
Add to Cart
Add to Cart
This item is not returnable. View return policy

used as a solid support for most sandwich ELISAs utilizing a mouse IgG capture antibody and a non-mouse IgG detection antibody, other applications include competitive ELISA, IgG isotyping, and hybridoma screening/selection.

Specifications

Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Surface Treatment EvenCoat Goat Anti-Mouse IgG
For Use With (Application) ELISA
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.