Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Polystyrene Microplates

Catalog No. DY991
Change view
Click to view available options
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
DY991 Black
DY990 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY991 Supplier R&D Systems™ Supplier No. DY991
Add to Cart
Add to Cart
This item is not returnable. View return policy

high-binding, flat-bottom polystyrene microplates

Specifications

Format Standard
Array 8 x 12
Color Black
Material Polystyrene
No. of Wells 96
Well Shape Flat Bottom (F-well)
For Use With (Application) ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.