Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Group 2
SDP

Catalog No. QC05 Shop All Novus Biologicals Products
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
QC05 Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC05 Supplier R&D Systems™ Supplier No. QC05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Control for Human IL-1 RII, IL-2 R alpha, IL-6 R alpha, TNF RI, TNF RII, VEGF R1, VEGF R2 Quantikine ELISAs

For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems' Quantikine™ kits. Controls are prepared in diluted porcine serum with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.