Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Random Primers

Catalog No. 48190011
Encompass_Preferred
Click to view available options
Quantity:
100 μL

Catalog No. 48-190-011

Supplier: Invitrogen™ 48190011

Only null left
Add to Cart
Add to Cart

Random Primers are oligodeoxyribonucleotides (mostly hexamers) used to prepare labeled DNA probes from templates for filter hybridization or in situ hybridization and to prime mRNAs with or without poly(A) for cDNA synthesis.

Random Primers are oligodeoxyribonucleotides (mostly hexamers) used to prepare labeled DNA probes from templates for filter hybridization or in situ hybridization and to prime mRNAs with or without poly(A) for cDNA synthesis. These primers are truly random and are suitable for DNA synthesis using Klenow fragments with DNA templates or for cDNA synthesis using reverse transcriptase with mRNA templates. To avoid storage buffer interference, Random Primers are supplied in a low-salt-concentration buffer.

Performance and quality testing: Incorporation of a radioactively labeled nucleotide is verified using control DNA in a Random Primers labeling reaction

Order Info

Shipping Conditions: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage • Random Primers in 3 mM Tris-HCl, 0.2 mM EDTA (100 μL at 3 μg/μL)

Store at –20°C.
Guaranteed stable for 6 months when properly stored.
Shipping Condition Approved for shipment on Wet or Dry Ice
Concentration 3 μg/μL
Primer Random
Quantity 100 μL
For Use With (Application) cDNA Synthesis
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.