Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANGAP1 Monoclonal Antibody (19C7)
GREENER_CHOICE

Catalog No. 330800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-080-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-080-0 Supplier Invitrogen™ Supplier No. 330800
  • $552.00
    $414.65 / Each of 1

    Save $137.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

RANGAP1 Monoclonal Antibody for Western Blot, ICC/IF, ELISA

Ran is a small signaling GTPase that is involved in nucleocytoplasmic transport. Two additional functions of animal Ran in the formation of spindle asters and the reassembly of the nuclear envelope in mitotic cells have been recently reported. In contrast to Ras or Rho, Ran is not associated with membranes. Instead, the spatial sequestering of its accessory proteins, the Ran GTPase-activating protein RanGAP and the nucleotide exchange factor RCC1, appears to define the local concentration of RanGTP vs. RanGDP involved in signaling. Mammalian RanGAP is bound to the nuclear pore by a mechanism involving the attachment of small ubiquitin-related modifier protein (SUMO) to its C terminus and the subsequent binding of the SUMOylated domain to the nucleoporin Nup358.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANGAP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 19C7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene RANGAP1
Gene Accession No. P46060, P46061
Gene Alias C79654; fug1; KIAA1835; MGC20266; mKIAA1835; RAN GTPase activating protein 1; RAN GTPase activating protein 1 a; Ran GTPase activating protein 1 b; Ran GTPase activating protein 1 L homeolog; Ran GTPase activating protein 1 S homeolog; ran GTPase-activating protein 1; RANGAP; RanGAP1; rangap1.L; rangap1.S; rangap1a; rangap1-a; rangap1b; rangap1-b; rna1p; SD; segregation distorter homolog; segregation distortion; XELAEV_18023679mg; XELAEV_18025680mg
Gene Symbols RANGAP1
Host Species Mouse
Immunogen A fusion protein consisting of the first 203 amino-terminal residues of the mouse RanGAP-1 protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19387, 362965, 5905
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.