Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Ranger™ Dielectric Pacs Boots

Catalog No. 19024967
Change view
Click to view available options
Size:
8
9
10
11
12
13
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-024967 10
19-024965 8
19-024966 9
19-024968 11
19-024969 12
19-024970 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-024967 Supplier Honeywell™ Supplier No. R6130RDM100
This item is not returnable. View return policy

Over-the-sock electrical hazard boots

  • Premium-grade rubber construction with safety yellow trim
  • Special ozone-resistant rubber compound
  • Flexible, hand-layered construction
  • Durable nylon linings for easy on/off
  • Ultra-composite shanks reinforce construction and support the feet
  • Height: 12 (30.5cm)

Not for use near explosives or where conductive footwear is required.

Compliance

Steel Toe meets ASTM F2413-11 M I/75 C/75 and EH

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM F2413-11 M I/75 C/75 EH
Color Red, Yellow
Grade Premium grade
Height (English) 12 in.
Includes Safety yellow trim
Liner Material Nylon
Material Rubber
Shank Material Ultra composite
Size 10
Sole Features Flexible, hand-layered
Toe Material Steel
Resistant To Ozone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.