Learn More
Honeywell™ Ranger™ Hi-Pac Work Boots
Description
- Hand-layered construction is dipped and then vulcanized to ensure waterproof protection
- Commercial grade rubber compound prevents ozone deterioration for longer life footwear
- Heavy Trac Tread™ sole and heel and knurled protective overlays are tough and durable
- Durable and comfortable fabric linings
- Steel toe and steel midsole
- Black
- Men's sizes: 6 to 15
Compliance
Meets ASTM™ F2413-11 M I/75 C/75 PR.
Specifications
Specifications
| Color | Black |
| Height (English) | 16 in. |
| Material | Rubber |
| Size | 15 |