Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANK (CD265) Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-200129341
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119387 200 μL
PIPA5122123 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119387 Supplier Invitrogen™ Supplier No. PA5119387
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RANK (CD265) Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANK (CD265)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TNFRSF11A
Gene Accession No. O35305, Q9Y6Q6
Gene Alias CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Symbols TNFRSF11A
Host Species Rabbit
Immunogen Recombinant protein Receptor Activator Of Nuclear Factor Kappa B. The antigen corresponds to amino acid range 330-615 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21934, 8792
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.