Learn More
TARGETMOL CHEMICALS INC Rivastigmine 200MG
Supplier: TARGETMOL CHEMICALS INC T0229200MG
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Rivastigmine (Exelon) is a Cholinesterase Inhibitor, used for therapy of Alzheimer disease. Purity 99.85%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.