Learn More
Cayman Chemical Rapamycin, 53123-88-9, 1 mg
SDP
Supplier:  Cayman Chemical 133461
Encompass_Preferred
Molecular Formula C51H79NO13, Purity ≥95%, Formula Weight 914.2, Synonyms: Sirolimus; An immunosuppressant used to prevent transplant rejection; potent inhibitor of IL-2 activation of lymphocytes (IC50 = 5 pM); specifically interacts with FKBP12 to form a complex which inhibits mTORC1.
Catalog No. NC0814569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.