Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rapamycin

Catalog No. PHZ1235
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHZ1235 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHZ1235 Supplier Invitrogen™ Supplier No. PHZ1235
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rapamycin

Rapamycin is a member of a family of macrolide immunosuppressants that bind to and inhibit the FK506 binding protein (FKBP) proline rotamase.

Format/Formulation: Lyophilized

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30 °C).
Gene ID (Entrez) 2475
Gene Symbol MTOR
Gene Alias FRAP, FRAP1, FRAP2, RAFT1, RAPT1
Protein mechanistic target of rapamycin (serine/threonine kinase)
Shipping Condition Wet Ice
Quantity 5 mg
Product Type Rapamycin
Molecular Weight (g/mol) 914.2
Form Solid
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.