Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rapamycin, Fisher BioReagents

Catalog No. BP29631 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29631 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29631

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53123-88-9
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.187
InChI Key QFJCIRLUMZQUOT-HPLJOQBZSA-N
PubChem CID 5284616
ChEBI CHEBI:9168
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC

Specifications

Type Rapamycin
Physical Form Powder/Solid
Packaging Clear Glass
Color White
Quantity 1 mg
Assay Percent Range > 98 %
Boiling Point 183°C
Merck Index 15, 8232
Formula Weight 914.2
Percent Purity >98%
Chemical Name or Material Rapamycin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.